Assay Details
| Gene Symbol: Wdr34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008498.2 | NP_001008498.2 | 8 - 9 | 1389 | 75 | |
| GenBank mRNA | AK014304.1 | - | 8 - 9 | 1388 | 75 | |
| BC085113.1 | - | 8 - 9 | 1345 | 30730999 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21101048 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21101050 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21101051 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21101049 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2157676 | AK014304.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2157676 | BC085113.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2157676 | NM_001008498.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 643216 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 445822 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635955 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 55071 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136978 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 693586 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506993 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 926155 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 645196 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 623333 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 172068 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 141101 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2157676 | AK014304.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2157676 | BC085113.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2157676 | NM_001008498.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21101060 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04469843 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00333548 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04469884 | - | - |
| Whole Mouse Genome G4122A | A_51_P438461 | AK014304.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P438461 | BC085113.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P438461 | NM_001008498.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04469884 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01438279 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01438280 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21101049 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21101050 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21101048 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21101051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21101018 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21101044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21101064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01955491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01397450 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01265294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01397452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21101032 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21101076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01438284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21101030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01596853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21101024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21101037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01719588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01955492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21101018 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01265296 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01541969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01265299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21101033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01330810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01397451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21101049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21101054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01801888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01541970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21101046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21101055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01876281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21101045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01397449 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21101026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01351255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01444366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21101057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01596852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21101075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02205198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21101052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21101067 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10481390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481384 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481392 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481385 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481387 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481386 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481391 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481388 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481389 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429174_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10481383 | - | - |
| Mouse Genome 430 2.0 Array | 1429174_at | - | - |
| Mouse Genome 430 2.0 Array | 1429174_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71820_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71820_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200027467.mm.1 | NM_001008498.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200027467.mm.1 | NM_001008498.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200027467.mm.1 | TC0200003260.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200027466.mm.1 | NM_001008498.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200027466.mm.1 | NM_001008498.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200027466.mm.1 | TC0200003260.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429174_at | - | - |
| Mouse Expression Set 430 | 1429174_PM_at | - | - |
| Murine Genome U74v2 Set | 168356_at | - | - |
| Murine Genome U74v2 Set | 168356_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429174_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429174_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 586736 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1233784 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1233784 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1233784 | - | - |

Wdr34
Genomic Map