Assay Details
| Gene Symbol: Dolpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290508.1 | NP_001277437.1 | 3 - 4 | 567 | 74 | |
| NM_001290509.1 | NP_001277438.1 | 2 - 3 | 360 | 74 | ||
| NM_020329.4 | NP_065062.1 | 2 - 3 | 360 | 74 | ||
| GenBank mRNA | AB030189.1 | - | 2 - 3 | 200 | 74 | |
| AK002544.1 | - | 3 - 4 | 406 | 74 | ||
| AK044141.1 | - | 2 - 3 | 199 | 74 | ||
| AK053923.1 | - | 2 - 3 | 189 | 74 | ||
| AK088065.1 | - | 2 - 3 | 204 | 74 | ||
| BB853200.1 | - | 2 - 3 | 363 | 74 | ||
| BC026544.1 | - | 2 - 3 | 184 | 4975127 | 74 | |
| BY133699.1 | - | 2 - 3 | 285 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002544.1, NM_001290508.1 transcripts.
Set Membership: |
|

Dolpp1
Genomic Map