Assay Details
| Gene Symbol: Dolpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290508.1 | NP_001277437.1 | 5 - 6 | 753 | 70 | |
| NM_001290509.1 | NP_001277438.1 | 4 - 5 | 546 | 70 | ||
| NM_020329.4 | NP_065062.1 | 4 - 5 | 546 | 70 | ||
| GenBank mRNA | AB030189.1 | - | 4 - 5 | 386 | 70 | |
| AK002544.1 | - | 5 - 6 | 592 | 70 | ||
| AK044141.1 | - | 4 - 5 | 385 | 70 | ||
| AK053923.1 | - | 4 - 5 | 375 | 70 | ||
| AK088065.1 | - | 4 - 5 | 390 | 70 | ||
| BC026544.1 | - | 4 - 5 | 370 | 4975127 | 70 | |
More Information
Set Membership: |
|

Dolpp1
Genomic Map