Assay Details
| Gene Symbol: Dolpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290508.1 | NP_001277437.1 | 7 - 8 | 985 | 62 | |
| NM_020329.4 | NP_065062.1 | 6 - 7 | 778 | 62 | ||
| GenBank mRNA | AB030189.1 | - | 6 - 7 | 618 | 62 | |
| AK002544.1 | - | 7 - 8 | 824 | 62 | ||
| AK010797.1 | - | 2 - 3 | 215 | 62 | ||
| AK044141.1 | - | 6 - 7 | 617 | 62 | ||
| AK053923.1 | - | 6 - 7 | 607 | 62 | ||
| BC026544.1 | - | 6 - 7 | 602 | 4975127 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dolpp1
Genomic Map