Assay Details
| Gene Symbol: Cercam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207298.2 | NP_997181.1 | 11 - 12 | 1564 | 66 | |
| GenBank mRNA | AK155384.1 | - | 11 - 12 | 1560 | 66 | |
| BC052176.1 | - | 11 - 12 | 1527 | 66 | ||
| BC138847.1 | - | 11 - 12 | 1543 | 8861869 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21099934 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21099936 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21099935 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21099938 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P559545 | BC138847.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P559545 | AK155384.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P559545 | BC052176.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P559545 | NM_207298.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P559545 | BC138847.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P559545 | AK155384.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P559545 | BC052176.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P559545 | NM_207298.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 118976 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 177041 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333334 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40305 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 397786 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 388046 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 152810 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126333 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123678 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P559545 | BC138847.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P559545 | AK155384.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P559545 | BC052176.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P559545 | NM_207298.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P559545 | BC138847.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P559545 | AK155384.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P559545 | BC052176.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P559545 | NM_207298.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099950 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126333 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00333239 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123678 | - | - |
| Whole Mouse Genome G4122A | A_52_P559545 | BC138847.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P559545 | AK155384.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P559545 | BC052176.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P559545 | NM_207298.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P559545 | BC138847.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P559545 | AK155384.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P559545 | BC052176.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P559545 | NM_207298.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104398 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121136 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126333 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123678 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01454692 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01152009 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01152008 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01152013 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01151997 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21099938 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21099936 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21099934 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21099935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21100013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02276867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01194088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099978 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01889653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01763095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099971 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01186009 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02177497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01806375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01417855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01766760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01287391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099901 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01763092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01417857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01152009 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099953 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21100013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01835492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02177498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099910 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10470786 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470780 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470787 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470785 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470777 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470778 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470776 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470781 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470783 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481382 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470779 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470782 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470784 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435345_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10470775 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10481380 | - | - |
| Mouse Genome 430 2.0 Array | 1435345_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1432448_at | - | - |
| Mouse Genome 430 2.0 Array | 1432448_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435345_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 99151_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 99151_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200003813.mm.1 | NM_207298.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200003813.mm.1 | NM_207298.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200003813.mm.1 | TC0200000514.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200003814.mm.1 | NM_207298.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200003814.mm.1 | NM_207298.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200003814.mm.1 | TC0200000514.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435345_at | - | - |
| Mouse Expression Set 430 | 1435345_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435345_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435345_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 823615 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 780605 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000780605 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2671755 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2671755 | - | - |
| mouse-6 v1.1 (Illumina) | scl0099151.1_201-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 780605 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0099151.1_201-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 780605 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2671755 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2671755 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0099151.1_201-S | - | - |

Cercam
Genomic Map