Assay Details
| Gene Symbol: Cercam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207298.2 | NP_997181.1 | 1 - 2 | 230 | 72 | |
| GenBank mRNA | AK155384.1 | - | 1 - 2 | 225 | 72 | |
| BB621603.1 | - | 1 - 2 | 230 | 72 | ||
| BC052176.1 | - | 1 - 2 | 191 | 72 | ||
| BC138847.1 | - | 1 - 2 | 209 | 8861869 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cercam
Genomic Map