| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600004488.mm.1 |
NM_001172216.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004488.mm.1 |
NM_001172216.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004488.mm.1 |
NM_172727.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004488.mm.1 |
NM_172727.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004488.mm.1 |
TC0600000585.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600004490.mm.1 |
NM_001172216.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004490.mm.1 |
NM_001172216.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004490.mm.1 |
NM_172727.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004490.mm.1 |
NM_172727.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004490.mm.1 |
TC0600000585.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600004489.mm.1 |
NM_001172216.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004489.mm.1 |
NM_001172216.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004489.mm.1 |
NM_172727.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004489.mm.1 |
NM_172727.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004489.mm.1 |
TC0600000585.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600004492.mm.1 |
NM_001172216.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004492.mm.1 |
NM_001172216.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004492.mm.1 |
NM_172727.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004492.mm.1 |
NM_172727.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004492.mm.1 |
TC0600000585.mm.1 |
MTA TC |