Assay Details
| Gene Symbol: Fam221a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172216.1 | NP_001165687.1 | 2 - 3 | 403 | 70 | |
| NM_172727.3 | NP_766315.1 | 2 - 3 | 403 | 70 | ||
| GenBank mRNA | AK052323.1 | - | 2 - 3 | 317 | 70 | |
| AK140239.1 | - | 2 - 3 | 309 | 70 | ||
| BC055818.1 | - | 2 - 3 | 331 | 4487036 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam221a
Genomic Map