Assay Details
| Gene Symbol: Cdan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026891.2 | NP_081167.2 | 8 - 9 | 1402 | 80 | |
| GenBank mRNA | AK157271.1 | - | 8 - 9 | 1403 | 80 | |
| BC117555.1 | - | 6 - 7 | 687 | 40098039 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdan1
Genomic Map