Assay Details
| Gene Symbol: Apbb1ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019456.2 | NP_062329.2 | 1 - 2 | 193 | 102 | |
| GenBank mRNA | AK040084.1 | - | 1 - 2 | 128 | 102 | |
| AK041552.1 | - | 1 - 2 | 196 | 102 | ||
| AK042467.1 | - | 1 - 2 | 131 | 102 | ||
| AK079736.1 | - | 1 - 2 | 427 | 102 | ||
| AK162323.1 | - | 1 - 2 | 77 | 102 | ||
| BB638237.1 | - | 1 - 2 | 195 | 102 | ||
| BC023110.1 | - | 1 - 2 | 88 | 3707478 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041552.1, AK162323.1, BC023110.1, NM_019456.2 transcripts.
Set Membership: |
|

Apbb1ip
Genomic Map