Assay Details
| Gene Symbol: Cenpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007682.2 | NP_031708.2 | 1 | 1868 | 64 | |
| GenBank mRNA | AK158811.1 | - | 1 | 457 | 64 | |
| BC053333.1 | - | 1 | 1868 | 6399142 | 64 | |
| BC071269.1 | - | 1 | 1923 | 6333993 | 64 | |
| BC075733.1 | - | 1 | 1743 | 30286478 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21633701 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21633702 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21633704 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21633705 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21633700 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21633703 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139799 | BC075733.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139799 | BC053333.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139799 | BC071269.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139799 | NM_007682.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139799 | AK158811.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077213 | BC075733.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077213 | BC053333.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077213 | BC071269.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077213 | NM_007682.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077213 | BC006628.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 722966 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105604 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077213 | BC075733.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077213 | BC053333.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077213 | BC071269.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077213 | NM_007682.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077213 | BC006628.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077218 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139799 | BC075733.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139799 | BC053333.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139799 | BC071269.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139799 | NM_007682.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139799 | AK158811.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P262250 | BC075733.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P262250 | BC053333.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P262250 | BC071269.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P262250 | NM_007682.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P262250 | AK158811.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633724 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633720 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633699 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633702 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21633693 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105604 | - | - |
| Whole Mouse Genome G4122A | A_52_P262250 | BC075733.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P262250 | BC053333.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P262250 | BC071269.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P262250 | NM_007682.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P262250 | AK158811.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139799 | BC075733.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139799 | BC053333.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139799 | BC071269.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139799 | NM_007682.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139799 | AK158811.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105604 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01722930 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02111838 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02111843 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01527260 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02111840 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02111841 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02111846 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01565279 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01527270 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21633701 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21633703 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21633704 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21633702 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21633700 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21633705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21633709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21633714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01527260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01527266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21633690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21633708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21633712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21633717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21633687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21633693 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21633709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02094313 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21633703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21633696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01527262 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01527270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01527267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21633711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21633718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21633699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21633713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01360721 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21633695 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10487872 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487877 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487878 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487873 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487875 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487874 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487876 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426051_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435944_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10487871 | - | - |
| Mouse Genome 430 2.0 Array | 1426051_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1435944_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1426051_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1435944_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12616_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12616_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200039976.mm.1 | TC0200004650.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200039971.mm.1 | NM_007682.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200039971.mm.1 | TC0200004650.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200039973.mm.1 | NM_007682.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200039973.mm.1 | TC0200004650.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200039972.mm.1 | NM_007682.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200039972.mm.1 | TC0200004650.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200039975.mm.1 | NM_007682.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200039975.mm.1 | TC0200004650.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426051_PM_a_at | - | - |
| Mouse Expression Set 430 | 1435944_PM_s_at | - | - |
| Mouse Expression Set 430 | 1426051_a_at | - | - |
| Mouse Expression Set 430 | 1435944_s_at | - | - |
| Murine Genome U74v2 Set | 99486_at | - | - |
| Murine Genome U74v2 Set | 99486_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435944_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426051_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435944_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426051_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426051_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426051_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1435944_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1435944_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 456702 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2940154 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2940154 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002940154 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2631771 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2631771 | - | - |
| mouse-6 v1.1 (Illumina) | scl18647.1_62-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2940154 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18647.1_62-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2940154 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2631771 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2631771 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl18647.1_62-S | - | - |

Cenpb
Genomic Map