Assay Details
| Gene Symbol: Nlrp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033431.1 | NP_001028603.1 | 9 - 10 | 3278 | 77 | |
| GenBank mRNA | AK149454.1 | - | 2 - 3 | 319 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
release of cytoplasmic sequestered NF-kappaB
negative regulation of signal transduction
negative regulation of protein autophosphorylation
negative regulation of NF-kappaB transcription factor activity
dendritic cell migration
regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of MHC class I biosynthetic process
negative regulation of interleukin-6 biosynthetic process
negative regulation of Toll signaling pathway
negative regulation of cytokine secretion
negative regulation of interleukin-1 secretion
negative regulation of inflammatory response
negative regulation of ERK1 and ERK2 cascade
cellular response to cytokine stimulus
negative regulation of NIK/NF-kappaB signaling
nucleotide binding
protein binding
ATP binding
release of cytoplasmic sequestered NF-kappaB
negative regulation of signal transduction
negative regulation of protein autophosphorylation
negative regulation of NF-kappaB transcription factor activity
dendritic cell migration
regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of MHC class I biosynthetic process
negative regulation of interleukin-6 biosynthetic process
negative regulation of Toll signaling pathway
negative regulation of cytokine secretion
negative regulation of interleukin-1 secretion
negative regulation of inflammatory response
negative regulation of ERK1 and ERK2 cascade
cellular response to cytokine stimulus
negative regulation of NIK/NF-kappaB signaling
nucleotide binding
protein binding
ATP binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971378 | AY364010.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971378 | BC116252.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971378 | BC116253.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971378 | NM_001033431.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971373 | NM_001033431.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971378 | AY364010.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971378 | BC116252.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971378 | BC116253.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971378 | NM_001033431.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971373 | NM_001033431.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09575349 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09575345 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09575335 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09575326 | - | - |
| Whole Mouse Genome G4122A | A_52_P658945 | NM_001033431.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P658945 | AK149454.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P50529 | AY364010.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P50529 | BC116252.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P50529 | BC116253.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P50529 | NM_001033431.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P50529 | AK158158.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09575335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33005248 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1440921_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 378425_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 378425_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700019113.mm.1 | NM_001033431.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019113.mm.1 | NM_001033431.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019113.mm.1 | TC0700002124.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1440921_PM_at | - | - |
| Mouse Expression Set 430 | 1440921_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 818277 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1253264 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1243089 | - | - |

Nlrp12
Genomic Map