Assay Details
| Gene Symbol: Acp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007387.2 | NP_031413.1 | 1 - 2 | 135 | 65 | |
| GenBank mRNA | AK089401.1 | - | 1 - 2 | 126 | 65 | |
| AK128963.1 | - | 1 - 2 | 128 | 65 | ||
| AK133756.1 | - | 1 - 2 | 131 | 65 | ||
| AK154269.1 | - | 1 - 2 | 131 | 65 | ||
| AK170658.1 | - | 1 - 2 | 135 | 65 | ||
| BC013495.1 | - | 1 - 2 | 125 | 65 | ||
| BC023343.1 | - | 1 - 2 | 117 | 5136781 | 65 | |
| X57199.1 | - | 1 - 2 | 106 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acp2
Genomic Map