Assay Details
| Gene Symbol: Acp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007387.2 | NP_031413.1 | 3 - 4 | 316 | 70 | |
| GenBank mRNA | AK089401.1 | - | 3 - 4 | 307 | 70 | |
| AK128963.1 | - | 3 - 4 | 309 | 70 | ||
| AK133756.1 | - | 3 - 4 | 312 | 70 | ||
| AK154269.1 | - | 3 - 4 | 312 | 70 | ||
| AK170658.1 | - | 3 - 4 | 316 | 70 | ||
| BC013495.1 | - | 3 - 4 | 306 | 70 | ||
| BC023343.1 | - | 3 - 4 | 298 | 5136781 | 70 | |
| X57199.1 | - | 3 - 4 | 287 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acp2
Genomic Map