Assay Details
| Gene Symbol: Rars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025936.3 | NP_080212.2 | 10 - 11 | 1283 | 107 | |
| GenBank mRNA | AK011383.1 | - | 10 - 11 | 1261 | 107 | |
| AK076160.1 | - | 10 - 11 | 1258 | 107 | ||
| AK149856.1 | - | 10 - 11 | 1258 | 107 | ||
| AK151168.1 | - | 4 - 5 | 449 | 107 | ||
| AK151966.1 | - | 9 - 10 | 1081 | 107 | ||
| AK152108.1 | - | 10 - 11 | 1258 | 107 | ||
| AK160216.1 | - | 10 - 11 | 1258 | 107 | ||
| AK166097.1 | - | 10 - 11 | 1258 | 107 | ||
| BC020132.1 | - | 10 - 11 | 1241 | 3989952 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rars
Genomic Map