Assay Details
| Gene Symbol: Rars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025936.3 | NP_080212.2 | 11 - 12 | 1389 | 76 | |
| GenBank mRNA | AK011383.1 | - | 11 - 12 | 1367 | 76 | |
| AK076160.1 | - | 11 - 12 | 1364 | 76 | ||
| AK149856.1 | - | 11 - 12 | 1364 | 76 | ||
| AK151168.1 | - | 5 - 6 | 555 | 76 | ||
| AK151966.1 | - | 10 - 11 | 1187 | 76 | ||
| AK152108.1 | - | 11 - 12 | 1364 | 76 | ||
| AK166097.1 | - | 11 - 12 | 1364 | 76 | ||
| AK168194.1 | - | 11 - 12 | 1364 | 76 | ||
| BC020132.1 | - | 11 - 12 | 1347 | 3989952 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rars
Genomic Map