Assay Details
| Gene Symbol: Rars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025936.3 | NP_080212.2 | 9 - 10 | 1103 | 164 | |
| GenBank mRNA | AK011383.1 | - | 9 - 10 | 1081 | 164 | |
| AK076160.1 | - | 9 - 10 | 1078 | 164 | ||
| AK149856.1 | - | 9 - 10 | 1078 | 164 | ||
| AK151168.1 | - | 3 - 4 | 269 | 164 | ||
| AK151966.1 | - | 8 - 9 | 901 | 164 | ||
| AK152108.1 | - | 9 - 10 | 1078 | 164 | ||
| AK160216.1 | - | 9 - 10 | 1078 | 164 | ||
| AK166097.1 | - | 9 - 10 | 1078 | 164 | ||
| AK168194.1 | - | 9 - 10 | 1078 | 164 | ||
| BC020132.1 | - | 9 - 10 | 1061 | 3989952 | 164 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rars
Genomic Map