Assay Details
| Gene Symbol: Otub2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026580.4 | NP_080856.1 | 2 - 3 | 287 | 144 | |
| GenBank mRNA | AK006054.1 | - | 2 - 3 | 209 | 144 | |
| AK006346.1 | - | 2 - 3 | 239 | 144 | ||
| AK006469.1 | - | 2 - 3 | 236 | 144 | ||
| AK016449.1 | - | 2 - 3 | 271 | 144 | ||
| AK019830.1 | - | 2 - 3 | 264 | 144 | ||
| AK160505.1 | - | 2 - 3 | 266 | 144 | ||
| BC049551.1 | - | 2 - 3 | 270 | 6771956 | 144 | |
| BC052409.1 | - | 2 - 3 | 202 | 144 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006054.1, AK006346.1, AK006469.1, AK019830.1, AK160505.1, BC049551.1, NM_026580.4 transcripts.
Set Membership: |
|

Otub2
Genomic Map