Assay Details
| Gene Symbol: Otub2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177841.1 | NP_001171312.1 | 5 - 6 | 920 | 84 | |
| NM_026580.4 | NP_080856.1 | 4 - 5 | 494 | 84 | ||
| NR_033587.1 | 4 - 5 | 776 | 84 | |||
| GenBank mRNA | AK006054.1 | - | 4 - 5 | 416 | 84 | |
| AK006346.1 | - | 4 - 5 | 446 | 84 | ||
| AK006469.1 | - | 4 - 5 | 443 | 84 | ||
| AK008258.2 | - | 4 - 5 | 378 | 84 | ||
| AK016449.1 | - | 4 - 5 | 478 | 84 | ||
| AK019830.1 | - | 4 - 5 | 471 | 84 | ||
| AK158758.1 | - | 4 - 5 | 777 | 84 | ||
| AK160505.1 | - | 4 - 5 | 473 | 84 | ||
| BC049551.1 | - | 4 - 5 | 477 | 6771956 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK158758.1 transcripts.
Set Membership: |
|

Otub2
Genomic Map