Assay Details
| Gene Symbol: C8g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027062.2 | NP_081338.1 | 1 - 2 | 413 | 56 | |
| GenBank mRNA | BC019967.1 | - | 1 - 2 | 413 | 4023907 | 56 |
| BY090016.1 | - | 1 - 2 | 182 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

C8g
Genomic Map