Assay Details
| Gene Symbol: C8g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271777.1 | NP_001258706.1 | 5 - 6 | 613 | 88 | |
| NM_027062.2 | NP_081338.1 | 5 - 6 | 834 | 88 | ||
| GenBank mRNA | AK005962.1 | - | 5 - 6 | 617 | 88 | |
| BC019967.1 | - | 5 - 6 | 834 | 4023907 | 88 | |
More Information
Set Membership: |
|

C8g
Genomic Map