Assay Details
| Gene Symbol: C8g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271777.1 | NP_001258706.1 | 6 - 7 | 654 | 64 | |
| NM_027062.2 | NP_081338.1 | 6 - 7 | 875 | 64 | ||
| GenBank mRNA | AK005962.1 | - | 6 - 7 | 658 | 64 | |
| BC019967.1 | - | 6 - 7 | 875 | 4023907 | 64 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135725 | NM_027062.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135725 | BC019967.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2151556 | NM_027062.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2151556 | AK005962.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2151556 | BC019967.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 547266 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171113 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2151556 | NM_027062.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2151556 | AK005962.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2151556 | BC019967.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135725 | NM_027062.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135725 | BC019967.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072589 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072587 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072597 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072592 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171113 | - | - |
| Whole Mouse Genome G4122A | A_52_P370225 | NM_027062.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P370225 | AK005962.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P370225 | BC019967.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171113 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01959940 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01959918 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01505254 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01959923 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01959931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072598 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01959949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01959936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01959937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01959944 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072598 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01959931 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01959951 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10480756 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480758 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480752 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480755 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480757 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480754 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480759 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480753 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451625_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10480751 | - | - |
| Mouse Genome 430 2.0 Array | 1451625_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1451625_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69379_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69379_PM_at | - | - |
| Mouse Expression Set 430 | 1451625_a_at | - | - |
| Mouse Expression Set 430 | 1451625_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451625_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451625_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451625_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451625_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 839131 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2637180 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2637180 | - | - |

C8g
Genomic Map