| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200010995.mm.1 |
NM_001109690.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010995.mm.1 |
NM_001109690.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010995.mm.1 |
NM_001109691.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010995.mm.1 |
NM_001109691.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010995.mm.1 |
NM_138755.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010995.mm.1 |
NM_138755.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010995.mm.1 |
TC0200001396.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200010994.mm.1 |
NM_001109690.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010994.mm.1 |
NM_001109690.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010994.mm.1 |
NM_001109691.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010994.mm.1 |
NM_001109691.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010994.mm.1 |
NM_138755.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010994.mm.1 |
NM_138755.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010994.mm.1 |
TC0200001396.mm.1 |
MTA TC |