| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200017221.mm.1 |
NR_040619.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200017221.mm.1 |
NR_040619.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200017221.mm.1 |
TC0200002173.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200017220.mm.1 |
NR_040619.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200017220.mm.1 |
NR_040619.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200017220.mm.1 |
TC0200002173.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200017222.mm.1 |
NR_040619.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200017222.mm.1 |
NR_040619.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200017222.mm.1 |
TC0200002173.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200017223.mm.1 |
NR_040619.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200017223.mm.1 |
NR_040619.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200017223.mm.1 |
TC0200002173.mm.1 |
MTA TC |