Assay Details
| Gene Symbol: Crnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081200.1 | NP_001074669.1 | 2 - 3 | 203 | 70 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BB618328.1 | - | 203 | Chr. - - [-] on Build | 70 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2136606 | NM_001081200.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 850639 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795981 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 141951 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2136606 | NM_001081200.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334739 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02793500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02876680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03071642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02585446 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02585433 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334742 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02585430 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02449501 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334816 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334739 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02585423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03088301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334741 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334743 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02876685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334787 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10493913 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493915 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493914 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10493912 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2528650 | - | - |

Crnn
Genomic Map