Assay Details
| Gene Symbol: Nxt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161430.2 | NP_001154902.1 | 1 - 2 | 75 | 102 | |
| GenBank mRNA | BB857403.1 | - | 1 - 2 | 76 | 103 | |
| CA482101.1 | - | 1 - 2 | 111 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001161430.2 transcripts.
Set Membership: |
|

Nxt2
Genomic Map