Assay Details
| Gene Symbol: Nxt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161430.2 | NP_001154902.1 | 3 - 4 | 253 | 96 | |
| NM_001290532.1 | NP_001277461.1 | 3 - 4 | 338 | 96 | ||
| NM_001290533.1 | NP_001277462.1 | 2 - 3 | 344 | 96 | ||
| NM_172782.4 | NP_766370.2 | 2 - 3 | 290 | 96 | ||
| GenBank mRNA | AK032102.1 | - | 2 - 3 | 261 | 96 | |
| AK144349.1 | - | 2 - 3 | 274 | 96 | ||
| BB851047.1 | - | 2 - 3 | 292 | 96 | ||
| BB857403.1 | - | 3 - 4 | 254 | 96 | ||
| BC068166.1 | - | 2 - 3 | 243 | 30361776 | 96 | |
| BC094570.1 | - | 2 - 3 | 207 | 30361291 | 96 | |
| BC118503.1 | - | 2 - 3 | 218 | 40106658 | 96 | |
| CA482101.1 | - | 3 - 4 | 289 | 96 | ||
| CJ148018.1 | - | 3 - 4 | 339 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001290533.1 transcripts.
Set Membership: |
|

Nxt2
Genomic Map