Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100006924.mm.1 |
NM_030679.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006924.mm.1 |
NM_030679.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006924.mm.1 |
TC1100000854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006919.mm.1 |
NM_030679.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006919.mm.1 |
NM_030679.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006919.mm.1 |
TC1100000854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006925.mm.1 |
NM_030679.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006925.mm.1 |
NM_030679.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006925.mm.1 |
TC1100000854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006920.mm.1 |
NM_030679.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006920.mm.1 |
NM_030679.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006920.mm.1 |
TC1100000854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006922.mm.1 |
NM_030679.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006922.mm.1 |
NM_030679.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006922.mm.1 |
TC1100000854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006921.mm.1 |
NM_030679.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006921.mm.1 |
NM_030679.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006921.mm.1 |
TC1100000854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006923.mm.1 |
NM_030679.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006923.mm.1 |
NM_030679.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006923.mm.1 |
TC1100000854.mm.1 |
MTA TC |