| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100006929.mm.1 |
NM_030679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006929.mm.1 |
NM_030679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006929.mm.1 |
TC1100000854.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100006926.mm.1 |
NM_030679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006926.mm.1 |
NM_030679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006926.mm.1 |
TC1100000854.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100006931.mm.1 |
NM_030679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006931.mm.1 |
NM_030679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006931.mm.1 |
TC1100000854.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100006930.mm.1 |
NM_030679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006930.mm.1 |
NM_030679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006930.mm.1 |
TC1100000854.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100006927.mm.1 |
NM_030679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006927.mm.1 |
NM_030679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006927.mm.1 |
TC1100000854.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100006928.mm.1 |
NM_030679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006928.mm.1 |
NM_030679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006928.mm.1 |
TC1100000854.mm.1 |
MTA TC |