| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100006904.mm.1 |
NM_030679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006904.mm.1 |
NM_030679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006904.mm.1 |
TC1100000854.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100006900.mm.1 |
NM_030679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006900.mm.1 |
NM_030679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006900.mm.1 |
TC1100000854.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100006902.mm.1 |
NM_030679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006902.mm.1 |
NM_030679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006902.mm.1 |
TC1100000854.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100006897.mm.1 |
NM_030679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006897.mm.1 |
NM_030679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006897.mm.1 |
TC1100000854.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100006903.mm.1 |
NM_030679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006903.mm.1 |
NM_030679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006903.mm.1 |
TC1100000854.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100006899.mm.1 |
NM_030679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006899.mm.1 |
NM_030679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006899.mm.1 |
TC1100000854.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100006901.mm.1 |
NM_030679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006901.mm.1 |
NM_030679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006901.mm.1 |
TC1100000854.mm.1 |
MTA TC |