Assay Details
Gene Symbol: Mrps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK077219.1 | - | 1 - 2 | 63 | 138 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK077219.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
