Assay Details
Gene Symbol: Kcnab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010599.4 | NP_034729.3 | 8 - 9 | 1050 | 103 | |
GenBank mRNA | AK029750.1 | - | 7 - 8 | 639 | 103 | |
AK031806.1 | - | 8 - 9 | 1031 | 103 | ||
AK044102.1 | - | 8 - 9 | 1005 | 103 | ||
AK046194.1 | - | 8 - 9 | 1040 | 103 | ||
AK049316.1 | - | 8 - 9 | 1051 | 103 | ||
BC017518.1 | - | 8 - 9 | 976 | 4507029 | 103 | |
U65593.1 | - | 1 - 2 | 161 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
