Assay Details
| Gene Symbol: Mok | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011973.2 | NP_036103.1 | 10 - 11 | 1163 | 128 | |
| GenBank mRNA | AB022695.1 | - | 10 - 11 | 1163 | 128 | |
| BC087925.1 | - | 5 - 6 | 602 | 6532951 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mok
Genomic Map