| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200016282.mm.1 |
NM_001079883.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016282.mm.1 |
NM_001079883.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016282.mm.1 |
TC1200002339.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200016283.mm.1 |
NM_001079883.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016283.mm.1 |
NM_001079883.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016283.mm.1 |
NM_021399.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016283.mm.1 |
NM_021399.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016283.mm.1 |
TC1200002339.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200016284.mm.1 |
NM_001079883.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016284.mm.1 |
NM_001079883.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016284.mm.1 |
NM_001286343.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016284.mm.1 |
NM_021399.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016284.mm.1 |
NM_021399.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016284.mm.1 |
TC1200002339.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200016285.mm.1 |
NM_001079883.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016285.mm.1 |
NM_001079883.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016285.mm.1 |
NM_001286343.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016285.mm.1 |
NM_021399.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016285.mm.1 |
NM_021399.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016285.mm.1 |
TC1200002339.mm.1 |
MTA TC |