Assay Details
| Gene Symbol: Entpd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172117.5 | NP_742115.2 | 6 - 7 | 811 | 63 | |
| GenBank mRNA | BC038126.1 | - | 6 - 7 | 800 | 1348160 | 63 |
More Information
Set Membership: |
|

Entpd6
Genomic Map