Assay Details
| Gene Symbol: Entpd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172117.5 | NP_742115.2 | 8 - 9 | 978 | 74 | |
| GenBank mRNA | AK156685.1 | - | 7 - 8 | 893 | 74 | |
| BC028318.1 | - | 8 - 9 | 922 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Entpd6
Genomic Map