Assay Details
| Gene Symbol: Rpp38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013376.2 | NP_001013394.2 | 1 - 2 | 106 | 118 | |
| GenBank mRNA | AK087782.1 | - | 1 - 2 | 107 | 118 | |
| BC051448.1 | - | 1 - 2 | 81 | 30060460 | 118 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK087782.1, BC051448.1, NM_001013376.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20961829 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20961833 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20961828 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20961826 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20961827 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20961830 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506674 | BC051448.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506674 | NM_001013376.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506674 | AK049133.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506674 | AK087782.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506674 | AK139023.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506674 | BC051448.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506674 | NM_001013376.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506674 | AK049133.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506674 | AK087782.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506674 | AK139023.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 347123 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 796526 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158725 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506674 | BC051448.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506674 | NM_001013376.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506674 | AK049133.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506674 | AK087782.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506674 | AK139023.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506674 | BC051448.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506674 | NM_001013376.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506674 | AK049133.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506674 | AK087782.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506674 | AK139023.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506674 | BC051448.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506674 | NM_001013376.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506674 | AK049133.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506674 | AK087782.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506674 | AK139023.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506674 | BC051448.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506674 | NM_001013376.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506674 | AK049133.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506674 | AK087782.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506674 | AK139023.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961823 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961826 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961830 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20961825 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158725 | - | - |
| Whole Mouse Genome G4122A | A_51_P506674 | BC051448.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506674 | NM_001013376.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506674 | AK049133.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506674 | AK087782.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506674 | AK139023.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506674 | BC051448.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506674 | NM_001013376.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506674 | AK049133.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506674 | AK087782.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506674 | AK139023.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158725 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01158768 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01158774 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01158773 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01158757 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01158775 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20961827 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20961833 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20961830 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20961828 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20961829 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20961826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20961856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20961797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01158770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20961795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01992874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20961817 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20961816 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20961798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20961841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20961837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20961802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01158754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20961818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20961839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01992881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20961799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01158764 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01992888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01158774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01158777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20961803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01992866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01158757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20961836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01992861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01813115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20961793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01158755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20961822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20961808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01598344 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01158762 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01158773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01992872 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10479750 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10479751 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10479749 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200023598.mm.1 | NM_001013376.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200023598.mm.1 | NM_001013376.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200023598.mm.1 | TC0200002844.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200023600.mm.1 | NM_001013376.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200023600.mm.1 | NM_001013376.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200023600.mm.1 | TC0200002844.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200023599.mm.1 | NM_001013376.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200023599.mm.1 | NM_001013376.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200023599.mm.1 | TC0200002844.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 628786 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1213674 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1213674 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2675514 | - | - |

Rpp38
Genomic Map