Assay Details
| Gene Symbol: Zbtb26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199025.2 | NP_950190.2 | 1 - 2 | 432 | 103 | |
| GenBank mRNA | AK166126.1 | - | 1 - 2 | 483 | 103 | |
| AK173205.1 | - | 1 - 2 | 57 | 103 | ||
| BC060655.1 | - | 1 - 2 | 432 | 30355035 | 103 | |
| BC109349.1 | - | 1 - 2 | 415 | 40056733 | 103 | |
| BC109350.1 | - | 1 - 2 | 415 | 40056735 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC060655.1, BC109349.1, BC109350.1, NM_199025.2 transcripts.
Set Membership: |
|

Zbtb26
Genomic Map