Assay Details
| Gene Symbol: Lhx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083125.1 | NP_001076594.1 | 3 - 4 | 533 | 63 | |
| NM_001083126.1 | NP_001076595.1 | 2 - 3 | 343 | 63 | ||
| NM_001083127.1 | NP_001076596.1 | 2 - 3 | 343 | 63 | ||
| NM_008500.2 | NP_032526.2 | 3 - 4 | 533 | 63 | ||
| GenBank mRNA | AB031039.1 | - | 2 - 3 | 343 | 63 | |
| AB031040.1 | - | 2 - 3 | 343 | 63 | ||
| AK090339.2 | - | 3 - 4 | 381 | 63 | ||
| AK140987.1 | - | 3 - 4 | 534 | 63 | ||
| BC065077.1 | - | 2 - 3 | 277 | 6816328 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lhx6
Genomic Map