Assay Details
| Gene Symbol: Lhx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083125.1 | NP_001076594.1 | 5 - 6 | 881 | 93 | |
| NM_001083126.1 | NP_001076595.1 | 4 - 5 | 691 | 93 | ||
| NM_001083127.1 | NP_001076596.1 | 4 - 5 | 691 | 93 | ||
| NM_008500.2 | NP_032526.2 | 5 - 6 | 881 | 93 | ||
| GenBank mRNA | AK140987.1 | - | 5 - 6 | 883 | 93 | |
| BC065077.1 | - | 4 - 5 | 625 | 6816328 | 93 | |
More Information
Set Membership: |
|

Lhx6
Genomic Map