Assay Details
| Gene Symbol: Golga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133852.2 | NP_598613.2 | 2 - 3 | 264 | 80 | |
| GenBank mRNA | AK216015.1 | - | 1 - 2 | 92 | 80 | |
| AK220509.1 | - | 2 - 3 | 192 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Golga2
Genomic Map