Assay Details
| Gene Symbol: Ccdc183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029859.1 | NP_084135.1 | 6 - 7 | 740 | 79 | |
| GenBank mRNA | AK014983.1 | - | 4 - 5 | 1572 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc183
Genomic Map