Assay Details
| Gene Symbol: 4933433C11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028961.1 | NP_083237.1 | 3 - 4 | 430 | 77 | |
| GenBank mRNA | AK029738.1 | - | 3 - 4 | 388 | 77 | |
| BC132286.1 | - | 3 - 4 | 430 | 40130563 | 77 | |
| BC138006.1 | - | 3 - 4 | 430 | 8861025 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

4933433C11Rik
Genomic Map