| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200025623.mm.1 |
NM_001162485.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200025623.mm.1 |
NM_001162485.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200025623.mm.1 |
NM_178408.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200025623.mm.1 |
NM_178408.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200025623.mm.1 |
NR_027876.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200025623.mm.1 |
NR_027876.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200025623.mm.1 |
TC0200003122.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200025626.mm.1 |
NM_001162485.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200025626.mm.1 |
NM_001162485.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200025626.mm.1 |
NM_178408.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200025626.mm.1 |
NM_178408.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200025626.mm.1 |
TC0200003122.mm.1 |
MTA TC |