Assay Details
| Gene Symbol: Ccdc73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177600.4 | NP_808268.2 | 15 - 16 | 1562 | 148 | |
| GenBank mRNA | AK029862.1 | - | 7 - 8 | 769 | 148 | |
| BC095976.1 | - | 14 - 15 | 1308 | 148 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc73
Genomic Map