Assay Details
| Gene Symbol: Ccdc73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177600.4 | NP_808268.2 | 6 - 7 | 585 | 159 | |
| GenBank mRNA | BB615231.1 | - | 6 - 7 | 583 | 159 | |
| BC095976.1 | - | 5 - 6 | 331 | 159 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC095976.1 transcripts.
Set Membership: |
|

Ccdc73
Genomic Map