Assay Details
| Gene Symbol: Ccdc73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177600.4 | NP_808268.2 | 8 - 9 | 760 | 149 | |
| GenBank mRNA | BC095976.1 | - | 7 - 8 | 506 | 149 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc73
Genomic Map