Assay Details
| Gene Symbol: Cep152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081091.1 | NP_001074560.1 | 8 - 9 | 1188 | 63 | |
| GenBank mRNA | AK030816.1 | - | 3 - 4 | 432 | 63 | |
| AK041913.1 | - | 8 - 9 | 1174 | 63 | ||
| AK173060.1 | - | 8 - 9 | 1188 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cep152
Genomic Map