Assay Details
| Gene Symbol: Gpr158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004761.1 | NP_001004761.1 | 6 - 7 | 2197 | 94 | |
| GenBank mRNA | AB093287.1 | - | 6 - 7 | 1302 | 94 | |
| AK030530.1 | - | 6 - 7 | 2193 | 94 | ||
| AK083234.1 | - | 6 - 7 | 2223 | 94 | ||
| AK086256.1 | - | 6 - 7 | 2199 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr158
Genomic Map