Assay Details
| Gene Symbol: Gpr158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004761.1 | NP_001004761.1 | 8 - 9 | 2587 | 109 | |
| GenBank mRNA | AB093287.1 | - | 8 - 9 | 1692 | 109 | |
| AK083234.1 | - | 8 - 9 | 2613 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr158
Genomic Map